Learn More
Aqua Solutions Technicon Standard #3 0.75 gpL NaClO₃, 150 gpL NaCl, 160 gpL NaOH (1L)
SDP
Catalog No. NC9524039
Encompass_Preferred
Technicon Standard #3 0.75 gpL NaClO₃, 150 gpL NaCl, 160 gpL NaOH (1L)
Catalog No. NC9524039 Supplier Aqua Solutions Supplier No. VUL365
Technicon Standard #3 0.75 gpL NaClO₃, 150 gpL NaCl, 160 gpL NaOH (1L)