Learn More
Braintree Scientific Techniplast Diuresis Cage for rats up to 300g, stand req.
SDP
Catalog No. 501954058
Encompass
Techniplast Diuresis Cage Urine flows along the inside surface of the collection funnel and is directed into the urine collection tube. The separating grid prevents feces to pass through. A simple twist unlocks either urine tube. No need to dismantle cage or disturbing the animal. Entire lower section of cage can be easily removed. 3700D000 Diuresis cage for rats up to 300 g all included (stand to be ordered separately) 3701D000 Diuresis cage for rats over 300 g all included (stand to be ordered separately) **Sold for research purposes only and not for human use**
Catalog No. 50-195-4058 Supplier Braintree Scientific Supplier No. 3700D000
Techniplast Diuresis Cage Urine flows along the inside surface of the collection funnel and is directed into the urine collection tube. The separating grid prevents feces to pass through. A simple twist unlocks either urine tube. No need to dismantle cage or disturbing the animal. Entire lower section of cage can be easily removed. 3700D000 Diuresis cage for rats up to 300 g all included (stand to be ordered separately) 3701D000 Diuresis cage for rats over 300 g all included (stand to be ordered separately) **Sold for research purposes only and not for human use**