Learn More
Braintree Scientific Techniplast Metabolic Cage Support Stand, Stand Only
SDP
Catalog No. 501954067
Encompass
Single - cage stand (Packed flat) Dimensions: mm 310 x 310 x 340 height Nylon supports. Holds one rodent metabolic cage or diuresis cage. Easy to assemble following package instructions. Autoclavable. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4067 Supplier Braintree Scientific Supplier No. 3M01D100
Single - cage stand (Packed flat) Dimensions: mm 310 x 310 x 340 height Nylon supports. Holds one rodent metabolic cage or diuresis cage. Easy to assemble following package instructions. Autoclavable. **Sold for research purposes only and not for human use**