Learn More
Braintree Scientific Technocut? Plus Disposable Safety Scalpel #12, 50/case
SDP
Catalog No. 501955940
Encompass
Technocut Plus Disposable Safety Scalpels- The safety and performance you need. Fully extended blade enables clear line of sight. Top activation allows for ambidextrous use . Fully enclosed front enhances blade stability and security. Audible clicks help confirm proper blade position. Final lock for safe handling and disposal. Non-slip ergonomic handle with metric scale. Color coded slider button for easy blade size identification. Highest quality standards. Uses premium GLASSVAN blades. Computerized grinding and proprietary polishing processes for the blades ensure consistent, clean, sharp cutting edges. Scalpel blades produced from high quality, high durability Swiss stainless steel. Scalpels manufactured in ISO 13485 certified facilities. **Sold for research purposes only and not for human use**
Catalog No. 50-195-5940 Supplier Braintree Scientific Supplier No. SSS12CS
Technocut Plus Disposable Safety Scalpels- The safety and performance you need. Fully extended blade enables clear line of sight. Top activation allows for ambidextrous use . Fully enclosed front enhances blade stability and security. Audible clicks help confirm proper blade position. Final lock for safe handling and disposal. Non-slip ergonomic handle with metric scale. Color coded slider button for easy blade size identification. Highest quality standards. Uses premium GLASSVAN blades. Computerized grinding and proprietary polishing processes for the blades ensure consistent, clean, sharp cutting edges. Scalpel blades produced from high quality, high durability Swiss stainless steel. Scalpels manufactured in ISO 13485 certified facilities. **Sold for research purposes only and not for human use**