Learn More
Grainger Tripp Lite Multi-Device Charger, Steel, 32-Port, Desktop/Wall-Mount
Catalog No. 502787347
Encompass_Preferred
This locking mobile phone charging station provides fast charging, convenient syncing, secure storage, and device organization for up to 32 smartphones or small USB devices. Constructed from durable steel with a black powder-coated finish, this stationary cabinet is designed for office, commercial, retail, industrial, and healthcare environments.
  • Supports fast 2.4 A charging per port for up to 32 devices.
  • Includes a USB uplink port for wired syncing and supports wireless syncing.
  • Features a 3-point locking mechanism with two included keys for security.
  • Offers flexible mounting options for wall or desktop installation.
  • Includes flow-through ventilation and attachment points for optional cooling fans.
  • Provides overcurrent and overcharge protection plus EMI/RFI line noise filtering.
  • Ships with a detachable 10 ft. (3.05 m) power cord with a right-angle NEMA 5-15P plug.
Catalog No. 50-278-7347 Supplier Grainger Supplier No. 199EJ0
This locking mobile phone charging station provides fast charging, convenient syncing, secure storage, and device organization for up to 32 smartphones or small USB devices. Constructed from durable steel with a black powder-coated finish, this stationary cabinet is designed for office, commercial, retail, industrial, and healthcare environments.
  • Supports fast 2.4 A charging per port for up to 32 devices.
  • Includes a USB uplink port for wired syncing and supports wireless syncing.
  • Features a 3-point locking mechanism with two included keys for security.
  • Offers flexible mounting options for wall or desktop installation.
  • Includes flow-through ventilation and attachment points for optional cooling fans.
  • Provides overcurrent and overcharge protection plus EMI/RFI line noise filtering.
  • Ships with a detachable 10 ft. (3.05 m) power cord with a right-angle NEMA 5-15P plug.