Learn More
Grainger Tripp Lite Mobile Charging Cart, 32-Device, Steel, Black, 1 ea.
Catalog No. 502787811
Encompass_Preferred
This mobile charging cart provides secure storage, efficient cord management, and USB charging for up to 32 tablets, smartphones, and other USB-powered devices. Designed for use in schools, medical facilities, and professional office environments, this cart features a durable steel construction with a black powder-coated finish to protect equipment from theft, damage, and tampering.
  • Provides fast 2.4A USB charging and syncing for up to 32 mobile devices
  • Includes protective shelves with removable dividers to accommodate various device thicknesses
  • Equipped with a built-in circuit breaker and flow-through ventilation
  • Constructed from steel with locking doors and included keys
  • Features large swivel casters with non-marking wheels and a reversible wide-grip handle for mobility
  • Supports wired syncing via an integrated USB uplink port
Catalog No. 50-278-7811 Supplier Grainger Supplier No. 199EH1
This mobile charging cart provides secure storage, efficient cord management, and USB charging for up to 32 tablets, smartphones, and other USB-powered devices. Designed for use in schools, medical facilities, and professional office environments, this cart features a durable steel construction with a black powder-coated finish to protect equipment from theft, damage, and tampering.
  • Provides fast 2.4A USB charging and syncing for up to 32 mobile devices
  • Includes protective shelves with removable dividers to accommodate various device thicknesses
  • Equipped with a built-in circuit breaker and flow-through ventilation
  • Constructed from steel with locking doors and included keys
  • Features large swivel casters with non-marking wheels and a reversible wide-grip handle for mobility
  • Supports wired syncing via an integrated USB uplink port