Learn More
Diapharma Group, Inc. Technothrombin Thrombin Generation Assay (TGA), 3 x 16 tests
SDP
Supplier:  Diapharma Group, Inc. 5006010
Encompass
Technothrombin TGA is a thrombin generation assay (TGA) based on monitoring the formation of thrombin by means of a fluorogenic substrate upon activation of the coagulation cascade by tissue factor.
Catalog No. NC0557944