Learn More
Electron Microscopy Sciences 3040 - Mounting Medium, 1/EA
SDP
Catalog No. 502090557
Encompass_Preferred
Technovit 3040 yellow is a fast curing methyl methacrylate-based resin, whose chemical composition warrants a firm, durable bond between Technovit and the specimen. Technovit 3040 consists of two components–powder and liquid–allowing simple mixing, easy adherance to the specimen, and fast curing. For fixing the mount, a high viscous consistency is required(i.e. a mixing ratio of approximately 2-3 parts per volume powder: 1 part per volume liquid) has proven to be the most advantageous. Includes: 1 x 1000g Technovit 3040 powder, 1 x 500 ml Technovit 3040 liquid.
Catalog No. 50-209-0557 Supplier Electron Microscopy Sciences Supplier No. 1465210L
Technovit 3040 yellow is a fast curing methyl methacrylate-based resin, whose chemical composition warrants a firm, durable bond between Technovit and the specimen. Technovit 3040 consists of two components–powder and liquid–allowing simple mixing, easy adherance to the specimen, and fast curing. For fixing the mount, a high viscous consistency is required(i.e. a mixing ratio of approximately 2-3 parts per volume powder: 1 part per volume liquid) has proven to be the most advantageous. Includes: 1 x 1000g Technovit 3040 powder, 1 x 500 ml Technovit 3040 liquid.