Learn More
Electron Microscopy Sciences Technovit 8100 Embedding Kit
SDP
Catalog No. 50980432
Encompass_Preferred
Technovit Glycol Methacrylate embedding kits developed by Kulzer in Germany are the most convenient to use and offers the best embedding results. Kits give better results offering improved morphology and allows for 1 micron serial sectioning. Polymerization temperature never exceeds 40 degree C, which makes either kit a potential choice for enzyme histochemistry.
The initiator is prepackaged, eliminating messy handling of dry components, and by eliminating aromatic amines from the formation, the components are significantly less toxic. The consistent high quality of raw materials used to make the resin allows for a permanent clear colorless block and, most important, reproducible results
Catalog No. 50-980-432 Supplier Electron Microscopy Sciences Supplier No. 14654
Technovit Glycol Methacrylate embedding kits developed by Kulzer in Germany are the most convenient to use and offers the best embedding results. Kits give better results offering improved morphology and allows for 1 micron serial sectioning. Polymerization temperature never exceeds 40 degree C, which makes either kit a potential choice for enzyme histochemistry.
The initiator is prepackaged, eliminating messy handling of dry components, and by eliminating aromatic amines from the formation, the components are significantly less toxic. The consistent high quality of raw materials used to make the resin allows for a permanent clear colorless block and, most important, reproducible results