Learn More
Electron Microscopy Sciences Technovit Embedding Mold HistoForm Q
SDP
Catalog No. 501902145
Encompass_Preferred
An important prerequisite for easy-to-cut specimens is a mold with a special profile which will provide specimen blocks for easy application and simple cutting. This is achieved with Histoform S, an embedding mold composed of new material with a special shape.
All essential requirements such as material-saving volume, easy-to-cut shape, unlimited shelf life, easy removal of the embedded specimens because of the conical shape and the PTFE material, excellent temperature dissipation by way of refined steel bottom, and accurate and easy fixing of the specimen blocks byspecial grooves were achieved with Histoform S, in cooperation with renowned users.
Curing temperature measurements have confirmed that at this volume the combination of Teflon and refined steel will result in a considerably lower polymerization temperature. Cavities are 20mm wide x16mm high x10mm deep.
Catalog No. 50-190-2145 Supplier Electron Microscopy Sciences Supplier No. 1465430
An important prerequisite for easy-to-cut specimens is a mold with a special profile which will provide specimen blocks for easy application and simple cutting. This is achieved with Histoform S, an embedding mold composed of new material with a special shape.
All essential requirements such as material-saving volume, easy-to-cut shape, unlimited shelf life, easy removal of the embedded specimens because of the conical shape and the PTFE material, excellent temperature dissipation by way of refined steel bottom, and accurate and easy fixing of the specimen blocks byspecial grooves were achieved with Histoform S, in cooperation with renowned users.
Curing temperature measurements have confirmed that at this volume the combination of Teflon and refined steel will result in a considerably lower polymerization temperature. Cavities are 20mm wide x16mm high x10mm deep.