Learn More
Med Vet International Technovit Liquid, 240cc, J0061LA
Catalog No. 5000495388
Encompass_Preferred
This popular acrylic consists of a powder and liquid that, when mixed together, form a strong binding adhesive material. The liquid and powder are mixed together in a roughly 1:2 ratio. Until it hardens, it can be poured and shaped. After it hardens, it can be sawed, drilled, or filed. It has a number of suggested uses. Cattle: a block or boot is adhered to the bottom of the healthy or unaffected claw, for sole abscesses or foot rot. Horses: quarter cracks, any hoof defects. Small animals: replace the connecting rod and K-E clamps for external fixation devices.
Catalog No. 50-004-95388 Supplier Med Vet International Supplier No. JOR61LA
This popular acrylic consists of a powder and liquid that, when mixed together, form a strong binding adhesive material. The liquid and powder are mixed together in a roughly 1:2 ratio. Until it hardens, it can be poured and shaped. After it hardens, it can be sawed, drilled, or filed. It has a number of suggested uses. Cattle: a block or boot is adhered to the bottom of the healthy or unaffected claw, for sole abscesses or foot rot. Horses: quarter cracks, any hoof defects. Small animals: replace the connecting rod and K-E clamps for external fixation devices.