Learn More
Cayman Chemical TechtochrysIn 10mg
SDP
Catalog No. 502912431
Encompass_Preferred
A flavonoid with diverse biological activities; induces adipogenesis and glucose uptake in 3T3-L1 adipocytes at 12.5, 25, and 50 UG/ml; active against S. aureus, L. monocytogenes, B. subtilis, S. enterica, E. coli, V. cholera, K. pneumonia, S. dysenteriae, and P. aeruginosa (MICs = 24-64 UG/ml); scavenges peroxyl radicals in a cell-free assay at 1 and 10 µM
Catalog No. 50-291-2431 Supplier Cayman Chemical Supplier No. 3579110 MG
A flavonoid with diverse biological activities; induces adipogenesis and glucose uptake in 3T3-L1 adipocytes at 12.5, 25, and 50 UG/ml; active against S. aureus, L. monocytogenes, B. subtilis, S. enterica, E. coli, V. cholera, K. pneumonia, S. dysenteriae, and P. aeruginosa (MICs = 24-64 UG/ml); scavenges peroxyl radicals in a cell-free assay at 1 and 10 µM