Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse TECK (CCL25) ELISA Kit

Catalog No. EMCCL25X10
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EMCCL25X10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EMCCL25X10 Supplier Invitrogen™ Supplier No. EMCCL25X10
Only null left
Add to Cart
Add to Cart
Request a Quote

ELISA

Chemokines are likely to play an important role in regulating the trafficking of developing T cells within the thymus. Chemokine C-C thymus expressed chemokine (TECK), also designated chemokine ligand 25 (CCL25), small induc-ible cytokine A25, chemokine b-15 or CK b-15, is expressed predominantly in thymic dendritic cells, thymic epithelial cells and in the small intestine. TECK, a CCR9 ligand, has suppressive activity against immature subsets of myeloid progenitors which have been stimulated to proliferate by multiple growth factors. TECK delivers signals through CCR9, which is important for the nav-igation of developing thymocytes. Bone marrow pre-pro-B cells and cells capable of generating pro-B colonies in the presence of interleukin-7 and Flt-3 ligand migrate to TECK, a response lost in later stages of B cell development.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O35903
Assay Range 20.5 to 2000 pg/mL
Assay Sensitivity 15 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 20300
Gene Symbol CCL25
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
Storage Requirements 2°C to 8°C
Target TECK
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.