Learn More
MSC Teclock Dial Indicator & Base Kit, 0.001 in Graduation, Magnetic Base, 3 Piece Set
Catalog No. 5000532331
Encompass
The Teclock Dial Indicator & Base Kit is a 3-piece set designed for precise measurements. It features a dial indicator with a 0.001 in graduation and a magnetic base for stable positioning. This kit is suitable for various laboratory and industrial applications requiring accurate dimensional readings.
  • 0.001 in graduation
  • Magnetic base style
  • Includes point kit set, dial indicator, and magnetic base
  • Sold as a 3 piece set
Catalog No. 50-005-32331 Supplier MSC Supplier No. 33102609
The Teclock Dial Indicator & Base Kit is a 3-piece set designed for precise measurements. It features a dial indicator with a 0.001 in graduation and a magnetic base for stable positioning. This kit is suitable for various laboratory and industrial applications requiring accurate dimensional readings.
  • 0.001 in graduation
  • Magnetic base style
  • Includes point kit set, dial indicator, and magnetic base
  • Sold as a 3 piece set