Learn More
Tecniplast™ Metabolic Cage Collection Funnel
Catalog No. 01259108 Shop All Tecniplast USA Inc. Products
Change view
Click to view available options
Material:
PMP
Catalog No. Material
01-259-108 PMP
1 options
Catalog No. 01-259-108 Supplier Tecniplast™ Supplier No. 3700M012
Add to Cart
Add to Cart
Replacement collection funnel for model 3700M022 double chamber metabolic mice cage
  • Material: polymethylpentene
  • Size: 8 7/8 dia. top x 8-1/3 in.H
Specifications
Product Type Collection Funnel
Material PMP
For Use With (Equipment) Model 3700M022 double chamber metabolic Mice cage