Learn More
Tecniplast™ Stainless Steel Raised Cage Lids without Divider
Catalog No. 012885K Shop All Tecniplast USA Inc. Products
Change view
Click to view available options
For Use With (Equipment):
Cage size P
Catalog No. For Use With (Equipment)
01-288-5K Cage size P
1 options
Catalog No. 01-288-5K Supplier Tecniplast™ Supplier No. 2000P242
Add to Cart
Add to Cart
Secure and easty to handle
  • U-shaped hopper allows easy access to food and won't entrap food or contaminants
  • Cross-wire members prevent bending
  • 0.19cm wire spacing
  • Autoclavable
Specifications
Product Type Raised Cage Lid without Divider
Material Stainless Steel
For Use With (Application) Guinea Pigs
For Use With (Equipment) Cage size P
Spacing (Metric) 0.19 cm