Learn More
Selleck Chemical LLC Tecovirimat-5mg
SDP
Supplier:  Selleck Chemical LLC S33805MG
Encompass_Preferred
Tecovirimat is an antiviral that inhibits the egress of orthopoxviruses by targeting viral p37 protein orthologs. Tecovirimat is developed for the treatment of smallpox infection.
Catalog No. 50-238-9631