Learn More
Medchemexpress LLC Tecovirimat-d4 | 869572-92-9 | 98.95% | 380.35 | 5 MG
SDP
Catalog No. 5000360496
Encompass_Preferred
Tecovirimat-d4 is a deuterium-labelled Tecovirimat (HY-14805), an orally bioavailable and selective compound effective against orthopoxviruses including vaccinia, monkeypox, camelpox, cowpox, ectromelia (mousepox), smallpox, and variola viruses. It targets the orthopoxvirus protein VP37, which is essential for membrane envelopment of intracellular mature virus particles.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-60496 Supplier Medchemexpress LLC Supplier No. HY14805S5MG
Tecovirimat-d4 is a deuterium-labelled Tecovirimat (HY-14805), an orally bioavailable and selective compound effective against orthopoxviruses including vaccinia, monkeypox, camelpox, cowpox, ectromelia (mousepox), smallpox, and variola viruses. It targets the orthopoxvirus protein VP37, which is essential for membrane envelopment of intracellular mature virus particles.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS