Learn More
Medchemexpress LLC Tectorigenin 7-O-Xylosyl Glucoside | 231288-19-0 | 5 MG
SDP
Catalog No. 5000419785
Encompass_Preferred
Tectorigenin 7-O-Xylosyl Glucoside is a glycosidic isoflavone isolated from Pueraria thomsonii flower, appearing as a white to off-white solid. It is classified under Flavonoids, Isoflavones, Phenols, and Polyphenols, and is intended for research use only.
  • Isolated from Pueraria thomsonii flower
  • Appears as a white to off-white solid
  • Classified under Flavonoids, Isoflavones, Phenols, and Polyphenols
  • Intended for research use only
Catalog No. 50-004-19785 Supplier Medchemexpress LLC Supplier No. HYN41725MG
Tectorigenin 7-O-Xylosyl Glucoside is a glycosidic isoflavone isolated from Pueraria thomsonii flower, appearing as a white to off-white solid. It is classified under Flavonoids, Isoflavones, Phenols, and Polyphenols, and is intended for research use only.
  • Isolated from Pueraria thomsonii flower
  • Appears as a white to off-white solid
  • Classified under Flavonoids, Isoflavones, Phenols, and Polyphenols
  • Intended for research use only