Learn More
Cayman Chemical TectrIgenIn 10mg
SDP
Catalog No. 502909227
Encompass_Preferred
An isoflavonoid with diverse biological activities and active metabolite of tectoridin; scavenges hydroxyl and superoxide radicals in cell-free assays (IC50s = 87 and 46.62 UG/ml, respectively); inhibits the formation of TBARS in egg yolk (IC50 = 23 UG/ml); inhibits the production of PGE2 induced by TPA or thapsigargin in isolated rat peritoneal macrophages at 3, 10, or 30 µM; reduces H2O2-induced cell death in V79-4 cells at 10 UG/ml; prevents increases in the hepatic levels of MDA and blood levels of ALT and AST and decreases in hepatic SOD, catalase, and GPX activities in a rat model of CCl4-induced liver injury at 100 mg/kg per day
Catalog No. 50-290-9227 Supplier Cayman Chemical Supplier No. 3586010 MG
An isoflavonoid with diverse biological activities and active metabolite of tectoridin; scavenges hydroxyl and superoxide radicals in cell-free assays (IC50s = 87 and 46.62 UG/ml, respectively); inhibits the formation of TBARS in egg yolk (IC50 = 23 UG/ml); inhibits the production of PGE2 induced by TPA or thapsigargin in isolated rat peritoneal macrophages at 3, 10, or 30 µM; reduces H2O2-induced cell death in V79-4 cells at 10 UG/ml; prevents increases in the hepatic levels of MDA and blood levels of ALT and AST and decreases in hepatic SOD, catalase, and GPX activities in a rat model of CCl4-induced liver injury at 100 mg/kg per day