Learn More
Medchemexpress LLC HY-125269 1mg , TED-347 CAS:2378626-29-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1252691MG
Encompass_Preferred
HY-125269 1mg TED-347 CAS: 2378626-29-8 TED-347 is a potent, irreversible, covalent and allosteric inhibitor at YAP-TEAD protein-protein interaction with an EC50 of 5.9 μM for TEAD4⋅Yap1 protein-protein interaction. TED-347 specifically and covalently bonds with Cys-367 within the central pocket of TEAD4 with a Ki of 10.3 μM. TED-347 blocks TEAD transcriptional activity and has antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0006