Learn More
Selleck Chemical LLC TED-347 2mg
Supplier: Selleck Chemical LLC S89512MG
TED-347 is a potent, irreversible, covalent and allosteric inhibitor of the TEAD?Yap protein-protein interaction. TED-347 inhibits TEAD4?Yap1 protein-protein interaction with an apparent EC50 of 5.9 ?M. *For Research & Development use only. Product is not intended for drug, household, or human consumption.