Learn More
Ted Pella Inc PELCO Histo Brush
SDP
Catalog No. NC1809404
Encompass
Boar hair brush for waste section removal. An aid for cleaning the knife stage and working area on the microtome. Total length: 27cm (10-5/8") L. Brush tip: 3.2cm (1-1/4") W x 3.2cm (1-1/4") L.
Catalog No. NC1809404 Supplier Ted Pella Inc Supplier No. 11873
Boar hair brush for waste section removal. An aid for cleaning the knife stage and working area on the microtome. Total length: 27cm (10-5/8") L. Brush tip: 3.2cm (1-1/4") W x 3.2cm (1-1/4") L.