Learn More
Medchemexpress LLC Tedatioxetine hydrobromide | 960151-65-9 | 99.8% | 364.34 g/mol | C18H22BrNS | 25 MG
SDP
Catalog No. 5000245624
Encompass_Preferred
Tedatioxetine hydrobromide is the hydrobromide salt of tedatioxetine, a serotonin- and norepinephrine-preferring triple reuptake inhibitor that also antagonizes 5-HT2A, 5-HT2C, 5-HT3, and α1A-adrenergic receptors. Offered as a solid powder and as pre-made DMSO solutions, it is intended for in vitro and preclinical research applications.
  • High purity (99.8%).
  • Triple reuptake inhibition with multi-receptor antagonism.
  • Available as solid powder and 10 mM DMSO solution.
  • Multiple pack sizes for flexible dosing and study scales.
  • Suitable for in vitro and preclinical pharmacology studies.
Catalog No. 50-002-45624 Supplier Medchemexpress LLC Supplier No. HY10175525MG
Tedatioxetine hydrobromide is the hydrobromide salt of tedatioxetine, a serotonin- and norepinephrine-preferring triple reuptake inhibitor that also antagonizes 5-HT2A, 5-HT2C, 5-HT3, and α1A-adrenergic receptors. Offered as a solid powder and as pre-made DMSO solutions, it is intended for in vitro and preclinical research applications.
  • High purity (99.8%).
  • Triple reuptake inhibition with multi-receptor antagonism.
  • Available as solid powder and 10 mM DMSO solution.
  • Multiple pack sizes for flexible dosing and study scales.
  • Suitable for in vitro and preclinical pharmacology studies.