Learn More
Cayman Chemical Tedizolid-13C-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 2931763-72-1, 1 mg
SDP
Catalog No. 502949500
Encompass_Preferred
An internal standard for the quantification of tedizolid by GC- or LC-MS
Catalog No. 50-294-9500 Supplier Cayman Chemical Supplier No. 332901 MG
An internal standard for the quantification of tedizolid by GC- or LC-MS