Learn More
Medchemexpress LLC HY-14855 5mg , Tedizolid CAS:856866-72-3 Purity:>98%
SDP
Catalog No. 502260883
Encompass_Preferred
HY-14855 5mg Tedizolid CAS: 856866-72-3 Tedizolid (TR 700; Torezolid; DA-7157) is a novel oxazolidinone, acting through inhibition of bacterial protein synthesis by binding to 23S ribosomal RNA (rRNA) of the 50S subunit of the ribosome. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-0883 Supplier Medchemexpress LLC Supplier No. HY148555MG
HY-14855 5mg Tedizolid CAS: 856866-72-3 Tedizolid (TR 700; Torezolid; DA-7157) is a novel oxazolidinone, acting through inhibition of bacterial protein synthesis by binding to 23S ribosomal RNA (rRNA) of the 50S subunit of the ribosome. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.