Learn More
Cayman Chemical Tedizolid Phosphate, Purity: ≥98%, CAS: 856867-55-5, 10 mg
SDP
Catalog No. 502994937
Encompass_Preferred
A prodrug form of tedizolid; reduces vegetation titer in a rabbit model of aortic valve endocarditis caused by MRSA (15 mg/kg); reduces densities of methicillin-susceptible S. aureus and MRSA infection in a mouse model of catheter-related biofilm infection; increases survival in a rabbit model of MRSA necrotizing pneumonia
Catalog No. 50-299-4937 Supplier Cayman Chemical Supplier No. 2372910 MG
A prodrug form of tedizolid; reduces vegetation titer in a rabbit model of aortic valve endocarditis caused by MRSA (15 mg/kg); reduces densities of methicillin-susceptible S. aureus and MRSA infection in a mouse model of catheter-related biofilm infection; increases survival in a rabbit model of MRSA necrotizing pneumonia