Learn More
TARGETMOL CHEMICALS INC Tedizolid Phosphate 25MG
SDP
Catalog No. 502877836
Encompass_Preferred
Also available in 2 mg, 5 mg, 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Tedizolid Phosphate (TR-701FA) is an oxazolidinone-class antibiotic. Tedizolid phosphate is a phosphate ester prodrug of the active compound tedizolid. Purity 99.84%
Catalog No. 50-287-7836 Supplier TARGETMOL CHEMICALS INC Supplier No. T699825MG
Also available in 2 mg, 5 mg, 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Tedizolid Phosphate (TR-701FA) is an oxazolidinone-class antibiotic. Tedizolid phosphate is a phosphate ester prodrug of the active compound tedizolid. Purity 99.84%