Learn More
Medchemexpress LLC HY-14855B 5mg Medchemexpress, Tedizolid (phosphate) CAS:856867-55-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY14855B5MG
Encompass_Preferred
Medchemexpress, HY-14855B 5mg Tedizolid (phosphate) CAS:856867-55-5 Tedizolid phosphate (TR-701FA) is a novel oxazolidinone with activity against Gram-positive pathogens. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1650