Learn More
Selleck Chemical LLC Tedizolid Phosphate S4641-25mg
SDP
Catalog No. 503111533
Encompass_Preferred
Tedizolid (TR-701FA) is an oxazolidinone-class antibiotic Tedizolid phosphate is a phosphate ester prodrug of the active compound tedizolid
Catalog No. 50-311-1533 Supplier Selleck Chemical LLC Supplier No. S464125MG
Tedizolid (TR-701FA) is an oxazolidinone-class antibiotic Tedizolid phosphate is a phosphate ester prodrug of the active compound tedizolid