Learn More
Medchemexpress LLC Tedizolid phosphate | 856867-55-5 | 99.6% | 450.32 | 100 MG
SDP
Catalog No. 5000362369
Encompass_Preferred
Tedizolid phosphate (TR-701FA) is an MAO-A and MAO-B inhibitor that shows activity against Gram-positive bacteria. It is intended for research use only. In vitro studies demonstrate that Tedizolid phosphate (0.25 μg/mL) inhibits all 28 clinical isolates of PRSP, being 4-fold more potent than linezolid against PRSP. In vivo, it achieved a 100% survival rate in mice infected with PSSP type III at a minimum total daily dose of 10 mg/kg, with treated mice showing reduced lung inflammation and edema.
  • MAO-A and MAO-B inhibitor
  • Active against Gram-positive bacteria
  • 4-fold more potent than linezolid against PRSP in vitro
  • Achieves 100% survival rate in PSSP type III infected mice
  • Reduces lung inflammation and edema in treated mice
Catalog No. 50-003-62369 Supplier Medchemexpress LLC Supplier No. HY14855B100MG
Tedizolid phosphate (TR-701FA) is an MAO-A and MAO-B inhibitor that shows activity against Gram-positive bacteria. It is intended for research use only. In vitro studies demonstrate that Tedizolid phosphate (0.25 μg/mL) inhibits all 28 clinical isolates of PRSP, being 4-fold more potent than linezolid against PRSP. In vivo, it achieved a 100% survival rate in mice infected with PSSP type III at a minimum total daily dose of 10 mg/kg, with treated mice showing reduced lung inflammation and edema.
  • MAO-A and MAO-B inhibitor
  • Active against Gram-positive bacteria
  • 4-fold more potent than linezolid against PRSP in vitro
  • Achieves 100% survival rate in PSSP type III infected mice
  • Reduces lung inflammation and edema in treated mice