Learn More
Medchemexpress LLC Tedizolid phosphate | 856867-55-5 | MFCD28098176 | 99.7% | 450.32 | C17H16FN6O6P | 10 MG
SDP
Catalog No. 5000286950
Encompass_Preferred
Tedizolid phosphate is a research-grade oxazolidinone prodrug used in laboratory and preclinical studies for its antibacterial activity and monoamine oxidase inhibition. It is supplied as a solid in several weights and as a DMSO solution for biochemical and microbiological assays.
  • Research-grade oxazolidinone prodrug with activity against Gram-positive bacteria.
  • Inhibits monoamine oxidase A and B.
  • High purity (99.65%).
  • Available in multiple solid sizes and as a 10 mM DMSO solution.
  • Soluble in DMSO and water, suitable for biochemical assays.
Catalog No. 50-002-86950 Supplier Medchemexpress LLC Supplier No. HY14855B10MG
Tedizolid phosphate is a research-grade oxazolidinone prodrug used in laboratory and preclinical studies for its antibacterial activity and monoamine oxidase inhibition. It is supplied as a solid in several weights and as a DMSO solution for biochemical and microbiological assays.
  • Research-grade oxazolidinone prodrug with activity against Gram-positive bacteria.
  • Inhibits monoamine oxidase A and B.
  • High purity (99.65%).
  • Available in multiple solid sizes and as a 10 mM DMSO solution.
  • Soluble in DMSO and water, suitable for biochemical assays.