Learn More
SKC Inc Tedlar 10-liter Sample Bag with single polypropylene septum fitting, 10/pack
SDP
Catalog No. NC1364795
Encompass
Tedlar Air Sample Bags are made from classic DuPont film - the industry standard - with good stability for VOCs, sulfur compounds, carbon monoxide, carbon dioxide, methane, and sulfur hexafluoride. Use PTFE tubing to attach a dual stainless steel or single polypropylene fitting on the bag to a Grab Air or Pocket Pump TOUCH. DOES NOT ship to Alaska, Arizona, California, Colorado, Hawaii, Idaho, Nevada, Oregon, Utah, Washington. Wyoming”
Catalog No. NC1364795 Supplier SKC Inc Supplier No. 23210
Tedlar Air Sample Bags are made from classic DuPont film - the industry standard - with good stability for VOCs, sulfur compounds, carbon monoxide, carbon dioxide, methane, and sulfur hexafluoride. Use PTFE tubing to attach a dual stainless steel or single polypropylene fitting on the bag to a Grab Air or Pocket Pump TOUCH. DOES NOT ship to Alaska, Arizona, California, Colorado, Hawaii, Idaho, Nevada, Oregon, Utah, Washington. Wyoming”