Learn More
SKC Inc Tedlar 5-liter Sample Bag with dual stainless steel fittings, 10/pack
SDP
Catalog No. NC0824353
Encompass
Tedlar Air Sample Bags are made from classic DuPont film - the industry standard - with good stability for VOCs, sulfur compounds, carbon monoxide, carbon dioxide, methane, and sulfur hexafluoride. Use PTFE tubing to attach a dual stainless steel or single polypropylene fitting on the bag to a Grab Air or Pocket Pump TOUCH. DOES NOT ship to Alaska, Arizona, California, Colorado, Hawaii, Idaho, Nevada, Oregon, Utah, Washington. Wyoming”
Catalog No. NC0824353 Supplier SKC Inc Supplier No. 23105
Tedlar Air Sample Bags are made from classic DuPont film - the industry standard - with good stability for VOCs, sulfur compounds, carbon monoxide, carbon dioxide, methane, and sulfur hexafluoride. Use PTFE tubing to attach a dual stainless steel or single polypropylene fitting on the bag to a Grab Air or Pocket Pump TOUCH. DOES NOT ship to Alaska, Arizona, California, Colorado, Hawaii, Idaho, Nevada, Oregon, Utah, Washington. Wyoming”