Learn More
SKC Inc Tedlar Sample Bags, 0.5 Liter, Single Polypropylene Fitting
SDP
Catalog No. NC9611126
Encompass
SKC Tedlar 0.5 Liter Sample Bag with single polypropylene septum fitting. Fitting combines the hose/valve and the septum holder into one compact fitting, (pkg/10). DOES NOT ship to Alaska, Arizona, California, Colorado, Hawaii, Idaho, Nevada, Oregon, Utah, Washington. Wyoming”
Catalog No. NC9611126 Supplier SKC Inc Supplier No. 23202
SKC Tedlar 0.5 Liter Sample Bag with single polypropylene septum fitting. Fitting combines the hose/valve and the septum holder into one compact fitting, (pkg/10). DOES NOT ship to Alaska, Arizona, California, Colorado, Hawaii, Idaho, Nevada, Oregon, Utah, Washington. Wyoming”