Learn More
SKC Inc Tedlar Sample Bags, 1 Liter, Dual Stainless Steel Fittings
SDP
Catalog No. NC9003032
Encompass
Tedlar Gas Sample Bag, 1L, 7 in. x 7 in., w/ Dual Stainless Steel Fittings, the hose/valve for flushing, filling, and sealing the bag, and the septum with a syringe port for removing samples for analysis DOES NOT ship to Alaska, Arizona, California, Colorado, Hawaii, Idaho, Nevada, Oregon, Utah, Washington. Wyoming”
Catalog No. NC9003032 Supplier SKC Inc Supplier No. 23101
Tedlar Gas Sample Bag, 1L, 7 in. x 7 in., w/ Dual Stainless Steel Fittings, the hose/valve for flushing, filling, and sealing the bag, and the septum with a syringe port for removing samples for analysis DOES NOT ship to Alaska, Arizona, California, Colorado, Hawaii, Idaho, Nevada, Oregon, Utah, Washington. Wyoming”