Learn More
SKC Inc Tedlar Sample Bags, 5 Liters, Single Polypropylene Fitting
SDP
Catalog No. NC0331636
Encompass
SKC Tedlar 5 Liter Sample Bag with single polypropylene septum fitting. Fitting combines the hose/valve and the septum holder into one compact fitting, (pkg/10). DOES NOT ship to Alaska, Arizona, California, Colorado, Hawaii, Idaho, Nevada, Oregon, Utah, Washington. Wyoming”
Catalog No. NC0331636 Supplier SKC Inc Supplier No. 23205
SKC Tedlar 5 Liter Sample Bag with single polypropylene septum fitting. Fitting combines the hose/valve and the septum holder into one compact fitting, (pkg/10). DOES NOT ship to Alaska, Arizona, California, Colorado, Hawaii, Idaho, Nevada, Oregon, Utah, Washington. Wyoming”