Learn More
Carbosynth LLC. Teduglutide, 5mg, Bolar Exemption applies.
SDP
Supplier:  Carbosynth LLC. TED3880PI5MG
Encompass_Preferred
Synonyms: H-His-Gly-Asp-Gly-Ser-Phe-Ser-Asp-Glu-Met-Asn-Thr-Ile-Leu-Asp-Asn-Leu-Ala-Ala-Arg-Asp-Phe-Ile-Asn-Trp-Leu-Ile-Gln-Thr-Lys-Ile-Thr-Asp-OH, [Gly2]-GLP-2 (human) , HGDGSFSDEMNTILDNLAARDFINWLIQTKITD, Application: Glucagon-like Peptide-2 Analog Used to Treat Short Bowel Syndrome , Salt Form:, Trifluoroacetate , Storage Temperature: -20 °C, References: Jeppesen, P., et al.: Gut, 54, 1224 (2005)., Jeppesen, P.B., Therap. Adv. Gastroenterol., 5,159 (2012)., D. J. Drucker et al., American Journal of Physiology - Gastrointestinal and Liver Physiology, 276(1), G79 (1999).
Catalog No. 50-168-7199