Learn More
Selleck Chemical LLC Teduglutide-5mg
SDP
Catalog No. 502389840
Encompass_Preferred
Teduglutide (ALX-0600, Gattex, Revestive, TAK 633) is an analogue of human glucagon-like peptide-2 (GLP-2) and binds to the GLP-2 receptors. Teduglutide prolongs the intestinotrophic properties of GLP-2 in animal models.
Catalog No. 50-238-9840 Supplier Selleck Chemical LLC Supplier No. S99355MG
Teduglutide (ALX-0600, Gattex, Revestive, TAK 633) is an analogue of human glucagon-like peptide-2 (GLP-2) and binds to the GLP-2 receptors. Teduglutide prolongs the intestinotrophic properties of GLP-2 in animal models.