Learn More
IDEX Health & Science Upchurch TEE/.020 THRU HOLES SS
Catalog No. NC1919395
Encompass_Preferred
Idex High-Pressure Tees, Diameter Outer: 0.5 mm, Stainless Steel, Diameter Inner: 0.02 in., For Use With: 1/16 in. O.D. Tubing, Swept Volume: 0.566 uL
Catalog No. NC1919395 Supplier IDEX Health & Science Upchurch Supplier No. U428
Idex High-Pressure Tees, Diameter Outer: 0.5 mm, Stainless Steel, Diameter Inner: 0.02 in., For Use With: 1/16 in. O.D. Tubing, Swept Volume: 0.566 uL