Learn More
Trajan Scientific VICI Valco Tee, Valco Fitting, 1/16" Tubing OD, 0.25mm Bore, Stainless Steel
Catalog No. 501090231
Encompass
VICI Valco Tee, VICI Valco Fitting, 1/16" Tubing OD, 0.25mm Bore, Stainless Steel. Made in the USA.
Catalog No. 50-109-0231 Supplier Trajan Scientific Supplier No. ZT1C
VICI Valco Tee, VICI Valco Fitting, 1/16" Tubing OD, 0.25mm Bore, Stainless Steel. Made in the USA.