Learn More
Trajan Scientific VICI Valco Tee Fingertight, PEEK, High Pressure, 1/16" Tubing OD, 0.50mm Bore, connect 3 lines, Cheminert
Catalog No. 501090435
Encompass
VICI Valco Tee, PEEK, High Pressure, 1/16" Tubing OD, 0.50mm Bore, connect 3 lines, Cheminert. Made in the USA.
Catalog No. 50-109-0435 Supplier Trajan Scientific Supplier No. ZT1MFPK
VICI Valco Tee, PEEK, High Pressure, 1/16" Tubing OD, 0.50mm Bore, connect 3 lines, Cheminert. Made in the USA.