Learn More
Grainger Tee 3/4 x 3/4 x 1/2 in CTS PEX Wht
Catalog No. 502768588
Encompass_Preferred
The Grainger 3/4" x 3/4" x 1/2" CTS PEX Tee is a white PEX plastic tee fitting for use in residential plumbing applications. It has a maximum pressure rating of 100 PSI at 73°F and is NSF-pw certified for use with potable water. The tee fitting has 3/4" outlets designed for use with PEX tubing and provides a straight-through flow path with one perpendicular outlet. It is made in the USA and is lead free. This tee fitting allows you to branch off plumbing lines and is commonly used to connect supply lines.
Catalog No. 50-276-8588 Supplier Grainger Supplier No. 16T752
The Grainger 3/4" x 3/4" x 1/2" CTS PEX Tee is a white PEX plastic tee fitting for use in residential plumbing applications. It has a maximum pressure rating of 100 PSI at 73°F and is NSF-pw certified for use with potable water. The tee fitting has 3/4" outlets designed for use with PEX tubing and provides a straight-through flow path with one perpendicular outlet. It is made in the USA and is lead free. This tee fitting allows you to branch off plumbing lines and is commonly used to connect supply lines.