Learn More
AVANTOR MASTERFLEX (VWR LLC) TEE 3IN CLAMP
Catalog No. 502742514
Encompass
Masterflex Fitting, Polypropylene, Tee, Sanitary Clamp Union, 3IN Clamp, Avantor
Catalog No. 50-274-2514 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3180666
Masterflex Fitting, Polypropylene, Tee, Sanitary Clamp Union, 3IN Clamp, Avantor