Learn More
Instech Replacement tee for Instech automated blood sampler. 18ga, 3-way, conductive. For large animal sampling with Instech ABS1 or ABS2 samplers.
SDP
Catalog No. 5080986
Encompass
The Instech ABS2/T18C3 is a specialized tubing set designed for the ABS2 Automated Blood Sampler™ (ABS2™) specifically for large animal applications. This VAB™ compatible tubing set features an 18-gauge configuration to support higher flow rates. The design integrates a low-loss reservoir that prevents hemodilution by returning the leading edge of each sample to the animal, reducing blood waste. Integrated PinPorts™ maintain a closed, aseptic system for manual sampling or flushing without breaking the fluid line.
Features
  • Application: Automated blood sampling for large animal species
  • Compatibility: Specifically for ABS2™ systems and large animal VABs™
  • Design: Low-loss configuration to minimize blood waste
  • Access: Integrated PinPorts™ for aseptic manual access
  • Specifications: 18ga (0.031 inch) ID for increased flow capacity
Catalog No. 50-809-86 Supplier Instech Supplier No. ABS2/T18C3
The Instech ABS2/T18C3 is a specialized tubing set designed for the ABS2 Automated Blood Sampler™ (ABS2™) specifically for large animal applications. This VAB™ compatible tubing set features an 18-gauge configuration to support higher flow rates. The design integrates a low-loss reservoir that prevents hemodilution by returning the leading edge of each sample to the animal, reducing blood waste. Integrated PinPorts™ maintain a closed, aseptic system for manual sampling or flushing without breaking the fluid line.
Features
  • Application: Automated blood sampling for large animal species
  • Compatibility: Specifically for ABS2™ systems and large animal VABs™
  • Design: Low-loss configuration to minimize blood waste
  • Access: Integrated PinPorts™ for aseptic manual access
  • Specifications: 18ga (0.031 inch) ID for increased flow capacity