Learn More
IDEX Health & Science Upchurch Tee Assembly PEEK for 1/8" OD
Catalog No. NC2022403
Encompass_Preferred
This item has a minimum order quantity of 5 per supplier requirements. Tee Assembly PEEK for 1/8" OD, .062 thru hole
Catalog No. NC2022403 Supplier IDEX Health & Science Upchurch Supplier No. P800
This item has a minimum order quantity of 5 per supplier requirements. Tee Assembly PEEK for 1/8" OD, .062 thru hole