Learn More
Phenomenex Inc Tee Assembly, SS
Catalog No. 502399567 Shop All Phenomenex Products
Encompass_Preferred
Tee Assembly, SS joins one 1/16" tubing with two 360µm OD Tubing (MAX 15,000 psi/ 1034 bar)
Catalog No. 50-239-9567 Supplier Phenomenex Inc Supplier No. AN01032
Tee Assembly, SS joins one 1/16" tubing with two 360µm OD Tubing (MAX 15,000 psi/ 1034 bar)