Learn More
Cole-Parmer TEE ASSY HP P-728 1EA
Catalog No. NC1897158 Shop All Cole-Parmer Products
Encompass
Idex P-728 High-Pressure Tee Assembly, PEEK, 0.050" ID, 1/16" OD Tubing, 10-32 Coned; 1/EA
Catalog No. NC1897158 Supplier Cole-Parmer Supplier No. 0202455
Idex P-728 High-Pressure Tee Assembly, PEEK, 0.050" ID, 1/16" OD Tubing, 10-32 Coned; 1/EA