Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Nylon, Tee, Hose Barb Union, 1"" ID; 10/PK
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3061334
Encompass
Barbed tee fittings are designed for quick and easy assembly. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
Catalog No. 50-630-344