Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Polypropylene, Tee, Hose Barb Union, 1""ID; 10/PK
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3061375/DEL
Encompass
Barbed tee fittings are designed for quick and easy assembly. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
Catalog No. NC1559239